4-Methoxybenzoic anhydride, 98%
4-Methoxybenzoic anhydride, 98%
4-Methoxybenzoic anhydride, 98%
Thermo Scientific Chemicals

4-Methoxybenzoic anhydride, 98%

CAS: 794-94-5 | C16H14O5 | 286.283 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06180.06
also known as L06180-06
Price (USD)
66.65
Online Exclusive
73.60
Save 6.95 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
73.60
Save 6.95 (9%)
Each
Chemical Identifiers
CAS794-94-5
IUPAC Name4-methoxybenzoyl 4-methoxybenzoate
Molecular FormulaC16H14O5
InChI KeyYGMHIBLUWGDWKP-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)94.5-100.5?C
Appearance (Color)White
Assay (Titration ex Anhydride)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
4-Methoxybenzoic anhydride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoic anhydride is used as pharmaceutical intermediates.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Moisture Sensitive. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only
Ryuki Kakino; Sayaka Yasumi; Isao Shimizu; Akio Yamamoto. Synthesis of Unsymmetrical Ketones by Palladium-Catalyzed Cross-Coupling Reaction of Carboxylic Anhydrides with Organoboron Compounds. Bulletin of the Chemical Society of Japan.2002, 75, (1), 137-148.