Methyl isovalerate, 98%
Methyl isovalerate, 98%
Methyl isovalerate, 98%
Thermo Scientific Chemicals

Methyl isovalerate, 98%

CAS: 556-24-1 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L06241.18
also known as L06241-18
Price (USD)
46.60
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
46.60
Each
Chemical Identifiers
CAS556-24-1
IUPAC Namemethyl 3-methylbutanoate
Molecular FormulaC6H12O2
InChI KeyOQAGVSWESNCJJT-UHFFFAOYSA-N
SMILESCOC(=O)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3910-1.3950 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only