n-Hexyl benzoate, 98+%
n-Hexyl benzoate, 98+%
n-Hexyl benzoate, 98+%
Thermo Scientific Chemicals

n-Hexyl benzoate, 98+%

CAS: 6789-88-4 | C13H18O2 | 206.285 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L06293.14
also known as L06293-14
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.20
Save 5.55
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.20
Save 5.55
Each
Chemical Identifiers
CAS6789-88-4
IUPAC Namehexyl benzoate
Molecular FormulaC13H18O2
InChI KeyUUGLJVMIFJNVFH-UHFFFAOYSA-N
SMILESCCCCCCOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4910-1.4950 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only