3-Ethyl-1-pentyn-3-ol, 98%
Thermo Scientific Chemicals

3-Ethyl-1-pentyn-3-ol, 98%

CAS: 6285-06-9 | C7H12O | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06335.06
also known as L06335-06
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS6285-06-9
IUPAC Name3-ethylpent-1-yn-3-ol
Molecular FormulaC7H12O
InChI KeyPUNRPAWKFTXZIW-UHFFFAOYSA-N
SMILESCCC(O)(CC)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4360-1.4400 @ 20?C
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only
Lucien BarnesJr; L. J. Molinini. Determination of Acetylenic Hydrogen by Means of Concentrated Silver Solutions.Anal. Chem.1955,27 (6), 1025-1027.