Octadecyltriethoxysilane, 98%, n-isomer 85% min
Octadecyltriethoxysilane, 98%, n-isomer 85% min
Octadecyltriethoxysilane, 98%, n-isomer 85% min
Thermo Scientific Chemicals

Octadecyltriethoxysilane, 98%, n-isomer 85% min

CAS: 7399-00-0 | C24H52O3Si | 416.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L06366.18
also known as L06366-18
Price (USD)
170.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
170.00
Each
Chemical Identifiers
CAS7399-00-0
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥85.0% (major isomer)
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5% (sum of isomers)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only