1,7-Heptanediol, 95%
1,7-Heptanediol, 95%
1,7-Heptanediol, 95%
Thermo Scientific Chemicals

1,7-Heptanediol, 95%

CAS: 629-30-1 | C7H16O2 | 132.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06384.14
also known as L06384-14
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS629-30-1
IUPAC Nameheptane-1,7-diol
Molecular FormulaC7H16O2
InChI KeySXCBDZAEHILGLM-UHFFFAOYSA-N
SMILESOCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (Silylated GC)≥94.0%
Refractive Index1.4530-1.4590 @ 20?C
FormLiquid
1,7-Heptanediol is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Heptanediol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in water, slightly soluble in ether.

Notes
Incompatible with reducing agents, oxidizing agents, Acid chlorides, Acid anhydrides and chloroformates. Store away from acid chlorides and acid anhydrides.
RUO – Research Use Only

General References:

  1. P. Knauth; R. Sabbah. Energetics of intra- and intermolecular bonds in ω-alkanediols. III. Thermochemical study of 1,6-hexanediol, 1,7-heptanediol, 1,8-octanediol, 1,9-nonanediol, and 1,10-decanediol at 298.15 K.Canadian Journal of Chemistry.1990,68 (5), 731-734.
  2. Lynda J. Bruce and Andrew J. Daugulis. Solvent Selection Strategies for Extractive Biocatalysis.Biotechnology Progress.1991,7 116-124.