n-Octyl acetate, 98+%
n-Octyl acetate, 98+%
n-Octyl acetate, 98+%
n-Octyl acetate, 98+%
Thermo Scientific Chemicals

n-Octyl acetate, 98+%

CAS: 112-14-1 | C10H20O2 | 172.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L06405.30
also known as L06405-30
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.30
Save 9.65 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.30
Save 9.65 (15%)
Each
Add to cart
Chemical Identifiers
CAS112-14-1
IUPAC Nameoctyl acetate
Molecular FormulaC10H20O2
InChI KeyYLYBTZIQSIBWLI-UHFFFAOYSA-N
SMILESCCCCCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (unspecified)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only