3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole, 98+%
3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole, 98+%
3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole, 98+%
Thermo Scientific Chemicals

3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole, 98+%

CAS: 3314-35-0 | C9H11N3 | 161.208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L06476.09
also known as L06476-09
Price (USD)
53.65
Online Exclusive
59.60
Save 5.95 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS3314-35-0
IUPAC Name1-phenyl-4,5-dihydro-1H-pyrazol-3-amine
Molecular FormulaC9H11N3
InChI KeyQENUTIJJGGTTPE-UHFFFAOYSA-N
SMILESNC1=NN(CC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)165.5-171.5?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only