5-Methyl-2-nitrobenzoic acid, 98+%
5-Methyl-2-nitrobenzoic acid, 98+%
5-Methyl-2-nitrobenzoic acid, 98+%
Thermo Scientific Chemicals

5-Methyl-2-nitrobenzoic acid, 98+%

CAS: 3113-72-2 | C8H7NO4 | 181.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06594.06
also known as L06594-06
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.90
Save 7.25
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.90
Save 7.25
Each
Chemical Identifiers
CAS3113-72-2
IUPAC Name5-methyl-2-nitrobenzoic acid
Molecular FormulaC8H7NO4
InChI KeyQRRSIFNWHCKMSW-UHFFFAOYSA-N
SMILESCC1=CC=C(C(=C1)C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)131.0-138.0?C
Appearance (Color)White to pale cream to cream to yellow
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only