Benzyl cyanoacetate, 97%
Benzyl cyanoacetate, 97%
Benzyl cyanoacetate, 97%
Thermo Scientific Chemicals

Benzyl cyanoacetate, 97%

CAS: 14447-18-8 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L06644.14
also known as L06644-14
Price (USD)
75.65
Online Exclusive
83.90
Save 8.25 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
83.90
Save 8.25 (10%)
Each
Chemical Identifiers
CAS14447-18-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale cream to cream to yellow to pale orange
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Melting Point (DSC)23.0-33.0?C
Assay (GC)≥96.0%
Refractive Index1.5165-1.5215 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only