5-Ethyl-2-furaldehyde, 98%
5-Ethyl-2-furaldehyde, 98%
5-Ethyl-2-furaldehyde, 98%
Thermo Scientific Chemicals

5-Ethyl-2-furaldehyde, 98%

CAS: 23074-10-4 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L06659.03
also known as L06659-03
Price (USD)
40.65
Online Exclusive
44.90
Save 4.25 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
44.90
Save 4.25 (9%)
Each
Chemical Identifiers
CAS23074-10-4
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1.0%
Assay (GC)≥97.5%
Appearance (Color)Clear pale yellow to yellow to red to brown
FormLiquid
Refractive Index1.5205-1.5245 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only