1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
Thermo Scientific Chemicals

1,6-Hexanedithiol, 97%

CAS: 1191-43-1 | C6H14S2 | 150.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L06686.14
also known as L06686-14
Price (USD)
61.65
Online Exclusive
68.80
Save 7.15 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.80
Save 7.15 (10%)
Each
Chemical Identifiers
CAS1191-43-1
IUPAC Namehexane-1,6-dithiol
Molecular FormulaC6H14S2
InChI KeySRZXCOWFGPICGA-UHFFFAOYSA-N
SMILESSCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5105-1.5155 @ 20?C
Identification (FTIR)Conforms
Assay (GC)≥96.0%
FormLiquid
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

Solubility
Not miscible in water.

Notes
Incompatible materials are Bases, Oxidizing agents, Reducing agents, Alkali metals. Avoid Heat and direct sunlight.
RUO – Research Use Only

General References:

  1. E. Hutter; J. H. Fendler and D. Roy. Surface Plasmon Resonance Studies of Gold and Silver Nanoparticles Linked to Gold and Silver Substrates by 2-Aminoethanethiol and 1,6-Hexanedithiol.J. Phys. Chem. B.2001,105 (45), 11159-11168.
  2. T. Y. B. Leung; M. C. Gerstenberg; D. J. Lavrich and G. Scoles. 1,6-Hexanedithiol Monolayers on Au(111): A Multitechnique Structural Study.Langmuir.2000,16 (2), 549-561.