N-Phenylthiourea, 97%
N-Phenylthiourea, 97%
N-Phenylthiourea, 97%
Thermo Scientific Chemicals

N-Phenylthiourea, 97%

CAS: 103-85-5 | C7H8N2S | 152.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number L06690.09
Price (USD)
49.60
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
49.60
Each
Chemical Identifiers
CAS103-85-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormCrystals or crystalline powder or powder and/or granules
Assay (HPLC)≥96.0%
CommentMay contain darker particulates from manufacturing process. Do NOT impact on product quality.
Identification (FTIR)Conforms
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

Solubility
Soluble in hot water and alcohol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

General References:

  1. Mekarat, S.; Pakawatchai, C.; Saithong, S. (N-Phenylthiourea-κS)bis(triphenylphosphane-κP)silver(I) nitrate. Acta. Cryst. 2014, 70 (7), m281-m282.
  2. Farquhar, D. R.; Kovatch, K. J.; Palmer, J. N.; Shofer, F. S.; Adappa, N. D.; Cohen, N. A. Phenylthiocarbamide taste sensitivity is associated with sinonasal symptoms in healthy adults. Int. Forum. Allergy. Rhinol. 2015, 5 (2), 111-118.