1-Naphthylacetonitrile, 97%
1-Naphthylacetonitrile, 97%
1-Naphthylacetonitrile, 97%
1-Naphthylacetonitrile, 97%
Thermo Scientific Chemicals

1-Naphthylacetonitrile, 97%

CAS: 132-75-2 | C12H9N | 167.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L06696.22
also known as L06696-22
Price (USD)
110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
130.00
Save 19.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
130.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS132-75-2
IUPAC Name2-(naphthalen-1-yl)acetonitrile
Molecular FormulaC12H9N
InChI KeyOQRMWUNUKVUHQO-UHFFFAOYSA-N
SMILESN#CCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)31.0-40.0?C
Appearance (Color)White to pale cream to cream to yellow to brown
Refractive Index1.6175-1.6225 @ 20?C
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only