1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals
1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals
1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals
1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L06768.09
also known as L06768-09
10 g
Catalog number L06768.09
also known as L06768-09
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2426-07-5
IUPAC Name2-[4-(oxiran-2-yl)butyl]oxirane
Molecular FormulaC8H14O2
InChI KeyLFKLPJRVSHJZPL-UHFFFAOYNA-N
SMILESC(CCC1CO1)CC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4435-1.4485 @ 20°C (non-U.S. sourced material)
FormLiquid
CommentMaterial Sourced in the U.S. and in other countries
Assay (GC)≥96.0%
1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water (partly miscible), acetone and heptane.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hao Chen; Zhifeng Jia; Deyue Yan; Xinyuan Zhu. Thermo-responsive highly branched polyethers by proton-transfer polymerization of 1,2,7,8-diepoxyoctane and multiols. Macromolecular Chemistry and Physics. 2007, 208,(15), 1637-1645.
  2. R.D.Schwartz; C.J.McCoy. Enzymatic epoxidation: synthesis of 7,8-epoxy-1-octene, 1,2-7,8-diepoxyoctane, and 1,2-Epoxyoctane by Pseudomonas oleovorans. Appl. Environ. Microbiol. 1976, 31,(1), 78-82.