2,6-Diisopropylphenol, 97%
2,6-Diisopropylphenol, 97%
2,6-Diisopropylphenol, 97%
Thermo Scientific Chemicals

2,6-Diisopropylphenol, 97%

An anesthetic that exhibits direct activation of the GABA-A receptor | CAS: 2078-54-8 | C12H18O | 178.275 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L06841.22
also known as L06841-22
Price (USD)
83.65
Special offer
Online exclusive
Ends: 30-Jun-2026
98.60
Save 14.95 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 30-Jun-2026
98.60
Save 14.95 (15%)
Each
Chemical Identifiers
CAS2078-54-8
IUPAC Name2,6-bis(propan-2-yl)phenol
Molecular FormulaC12H18O
InChI KeyOLBCVFGFOZPWHH-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC(C(C)C)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 96.0%
Appearance (Color)Clear, pale yellow to orange-brown
Refractive Index1.5115 - 1.5165 @ 20 *C
Formliquid
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.

Solubility
Very slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. C.Frenkel; B.W.Urban. Human brain sodium channels as one of the molecular target sites for the new intravenous anaesthetic propofol (2,6-diisopropylphenol). European Journal of Pharmacology. 1991, 208,(1), 75-79.
  2. P.G.Murphy; D.S.Myers; M.J.Davies; N.R.Webster; J.G.Jones. The antioxidant potential of propofol (2,6-diisopropylphenol). British Journal of Anaesthesia. 1992, 68,(6), 613-618.