2-Amino-6-chlorobenzonitrile, 98%
2-Amino-6-chlorobenzonitrile, 98%
Thermo Scientific Chemicals

2-Amino-6-chlorobenzonitrile, 98%

CAS: 6575-11-7 | C7H5ClN2 | 152.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L06883.03
also known as L06883-03
Price (USD)
25.65
Online Exclusive
27.80
Save 2.15 (8%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
25.65
Online Exclusive
27.80
Save 2.15 (8%)
Each
Add to cart
Chemical Identifiers
CAS6575-11-7
IUPAC Name2-amino-6-chlorobenzonitrile
Molecular FormulaC7H5ClN2
InChI KeyMEJVTQKBWPYBFG-UHFFFAOYSA-N
SMILESNC1=CC=CC(Cl)=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown
Melting Point (clear melt)132.0-138.0?C
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only