Decanophenone, 98+%
Decanophenone, 98+%
Decanophenone, 98+%
Thermo Scientific Chemicals

Decanophenone, 98+%

CAS: 6048-82-4 | C16H24O | 232.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06923.14
also known as L06923-14
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS6048-82-4
IUPAC Name1-phenyldecan-1-one
Molecular FormulaC16H24O
InChI KeyQQXJNLYVPPBERR-UHFFFAOYSA-N
SMILESCCCCCCCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder or fused solid or waxy solid
Melting Point (clear melt)32.0-39.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only