Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Thermo Scientific Chemicals

Carbonyl cyanide 3-chlorophenylhydrazone, 98%

CAS: 555-60-2 | C9H5ClN4 | 204.617 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L06932.MC
also known as L06932-MC
Price (USD)
28.90
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
28.90
Each
Chemical Identifiers
CAS555-60-2
IUPAC NameN-(3-chlorophenyl)-1-cyanomethanecarbohydrazonoyl cyanide
Molecular FormulaC9H5ClN4
InChI KeyUGTJLJZQQFGTJD-UHFFFAOYSA-N
SMILESClC1=CC=CC(NN=C(C#N)C#N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to dark orange to brown
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.

Solubility
Insoluble in water. Soluble in DMSO (5 mg/ml), ethanol (1 mg/ml) and methanol (10 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. A.J.Verhoeven; B.P.Neve; H.Jansen. Secretion and apparent activation of human hepatic lipase requires proper oligosaccharide processing in the endoplasmic reticulum. Biochemical Journal. 1999, 337,(Pt 1), 133-140.
  2. G.Biswas; O.A.Adebanjo; B.D.Freedman; H.K.Anandatheerthavarada; C.Vijayasarathy; M.Zaidi; M.Kotlikoff; N.G.Avadhani. Retrograde Ca2+ signaling in C2C12 skeletal myocytes in response to mitochondrial genetic and metabolic stress: a novel mode of inter-organelle crosstalk. EMBO Journal. 1999, 18,(3), 522-533.
  3. Uncoupler of oxidative phosphorylation: Biochem. Biophys. Acta, 394 145 (1975); Biochemistry, 17, 2245 (1978).