3,3-Dimethyl-1,2-epoxybutane, 95%
3,3-Dimethyl-1,2-epoxybutane, 95%
3,3-Dimethyl-1,2-epoxybutane, 95%
Thermo Scientific Chemicals

3,3-Dimethyl-1,2-epoxybutane, 95%

CAS: 2245-30-9 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06951.06
also known as L06951-06
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS2245-30-9
IUPAC Name2-tert-butyloxirane
Molecular FormulaC6H12O
InChI KeyHEAYDCIZOFDHRM-UHFFFAOYNA-N
SMILESCC(C)(C)C1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3970-1.4030 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1,2-epoxybutane is used as pharm and intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only