Benzoyl isothiocyanate, 98%
Benzoyl isothiocyanate, 98%
Benzoyl isothiocyanate, 98%
Thermo Scientific Chemicals

Benzoyl isothiocyanate, 98%

CAS: 532-55-8 | C8H5NOS | 163.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06964.14
also known as L06964-14
Price (USD)
146.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
146.00
Each
Chemical Identifiers
CAS532-55-8
IUPAC Namebenzoyl isothiocyanate
Molecular FormulaC8H5NOS
InChI KeyCPEKAXYCDKETEN-UHFFFAOYSA-N
SMILESO=C(N=C=S)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.6355-1.6395 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only