N-Dodecylmethylamine, 98%
N-Dodecylmethylamine, 98%
N-Dodecylmethylamine, 98%
Thermo Scientific Chemicals

N-Dodecylmethylamine, 98%

CAS: 7311-30-0 | C13H29N | 199.382 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07025.03
also known as L07025-03
Price (USD)
42.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
42.10
Each
Chemical Identifiers
CAS7311-30-0
IUPAC Nametridecan-1-aminium chloride
Molecular FormulaC13H30ClN
InChI KeyXDRDMVYFQARLNH-UHFFFAOYSA-N
SMILES[Cl-].CCCCCCCCCCCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4385-1.4425 @ 20°C (non-U.S. sourced material)
Appearance (Color)Clear colorless
CommentMaterial sourced in the U.S. and in other countries
Proton NMRConforms to structure (U.S. sourced material)
View more
N-Dodecylmethylamine is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Dodecylmethylamine is used as intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Yongsoon Shin; Li-Qiong Wang; Glen E Fryxell; Greg J Exarhos. Hygroscopic growth of self-assembled layered surfactant molecules at the interface between air and organic salts.Journal of Colloid and Interface Science.2005,284 (1), 278-281.
  2. B L Kabacoff; M L Douglass; I E Rosenberg; L W Levan; J K Punwar; S F Vielhuber; R J Lechner. Formation of nitrosamines in non-ionic and anionic emulsions in the presence and absence of inhibitors.IARC scientific publications.1984, (57), 347-352..