2-(2-Aminophenyl)benzimidazole, 98%
2-(2-Aminophenyl)benzimidazole, 98%
2-(2-Aminophenyl)benzimidazole, 98%
2-(2-Aminophenyl)benzimidazole, 98%
Thermo Scientific Chemicals

2-(2-Aminophenyl)benzimidazole, 98%

CAS: 5805-39-0 | C13H11N3 | 209.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07034.06
also known as L07034-06
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.80
Save 7.15 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.80
Save 7.15 (15%)
Each
Add to cart
Chemical Identifiers
CAS5805-39-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Assay (HPLC)≥97.5%
Melting Point (clear melt)210-217?C
FormPowder
Identification (FTIR)Conforms
2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Swadeshmukul Santra; Sneh K.Dogra. Excited-state intramolecular proton transfer in 2-(2'-aminophenyl) benzimidazole. Chemical Physics. 1998, 226,(3), 285-296. 
  2. Pilar Souza; Jose A.Garcia-Vazquez; Jose R.Masaguer. Complexes of divalent nickel and copper with the Schiff base derived from 2-(2-aminophenyl)benzimidazole and benzaldehyde. Transition Metal Chemistry. 1984, 9,(8), 318-321.