1-Heptanethiol, 98%
1-Heptanethiol, 98%
1-Heptanethiol, 98%
Thermo Scientific Chemicals

1-Heptanethiol, 98%

CAS: 1639-09-4 | C7H16S | 132.265 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07079.22
also known as L07079-22
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS1639-09-4
IUPAC Nameheptane-1-thiol
Molecular FormulaC7H16S
InChI KeyVPIAKHNXCOTPAY-UHFFFAOYSA-N
SMILESCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4495-1.4535 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only