trans-1,2-Cyclohexanediol, 98%
trans-1,2-Cyclohexanediol, 98%
trans-1,2-Cyclohexanediol, 98%
trans-1,2-Cyclohexanediol, 98%
Thermo Scientific Chemicals

trans-1,2-Cyclohexanediol, 98%

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07088.22
also known as L07088-22
Price (USD)
202.65
Special offer
Online exclusive
Ends: 30-Jun-2026
238.00
Save 35.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
202.65
Special offer
Online exclusive
Ends: 30-Jun-2026
238.00
Save 35.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1460-57-7
IUPAC Name(1R,2R)-cyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-PHDIDXHHSA-N
SMILESO[C@@H]1CCCC[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)99.0-106.0?C
Appearance (Color)White to cream
Assay (GC)≥97.5%
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Gaetan Caron; Romas J.Kazlauskas. An optimized sequential kinetic resolution of trans-1,2-cyclohexanediol. J. Org. Chem. 1991, 56,(26), 7251-7256.
  2. Andreas Hartung; Mark A.Keane; Arno Kraft. Advantages of synthesizing trans-1,2-cyclohexanediol in a continuous flow microreactor over a standard glass apparatus. J. Org. Chem. 2007, 72,(26), 10235-10238.