2-(Allylthio)nicotinic acid, 98%
2-(Allylthio)nicotinic acid, 98%
2-(Allylthio)nicotinic acid, 98%
Thermo Scientific Chemicals

2-(Allylthio)nicotinic acid, 98%

CAS: 175135-25-8 | C9H9NO2S | 195.24 g/mol
Have Questions?
Catalog NumberQuantity
L07090.06
also known as L07090-06
5 g
Catalog number L07090.06
also known as L07090-06
Price (USD)
51.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
51.20
Each
Chemical Identifiers
CAS175135-25-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder or fused solid or needles
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only