Diphenyl phosphoramidate, 97%
Diphenyl phosphoramidate, 97%
Diphenyl phosphoramidate, 97%
Thermo Scientific Chemicals

Diphenyl phosphoramidate, 97%

CAS: 2015-56-7 | C12H12NO3P | 249.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07155.06
also known as L07155-06
Price (USD)
62.65
Online Exclusive
69.90
Save 7.25 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
69.90
Save 7.25 (10%)
Each
Chemical Identifiers
CAS2015-56-7
IUPAC Name{[amino(phenoxy)phosphoryl]oxy}benzene
Molecular FormulaC12H12NO3P
InChI KeyQWMUDOFWQWBHFI-UHFFFAOYSA-N
SMILESNP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)145.0-154.0?C
Assay (HPLC)≥96.0%
Appearance (Color)White
Diphenyl phosphoramidate is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphoramidate is used as primary and secondary intermediates.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only