1-Octen-3-one, 97%, stab. with 0.1% BHA
1-Octen-3-one, 97%, stab. with 0.1% BHA
1-Octen-3-one, 97%, stab. with 0.1% BHA
Thermo Scientific Chemicals

1-Octen-3-one, 97%, stab. with 0.1% BHA

CAS: 4312-99-6 | C8H14O | 126.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07189.06
also known as L07189-06
Price (USD)
200.65
Online Exclusive
223.00
Save 22.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
200.65
Online Exclusive
223.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS4312-99-6
IUPAC Nameoct-1-en-3-one
Molecular FormulaC8H14O
InChI KeyKLTVSWGXIAYTHO-UHFFFAOYSA-N
SMILESCCCCCC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.4320-1.4385 @ 20°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only