1-Octanethiol, 98%
1-Octanethiol, 98%
1-Octanethiol, 98%
Thermo Scientific Chemicals

1-Octanethiol, 98%

CAS: 111-88-6 | C8H18S | 146.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L07195.AE
also known as L07195-AE
Price (USD)
36.70
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
36.70
Each
Chemical Identifiers
CAS111-88-6
IUPAC Nameoctane-1-thiol
Molecular FormulaC8H18S
InChI KeyKZCOBXFFBQJQHH-UHFFFAOYSA-N
SMILESCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4525-1.4570 @ 20°C
FormLiquid
1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanethiol can be used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Qin Jia Cai; Mary B.Chan-Park; Zhi Song Lu; Chang Ming Li; Beng S.Ong. Bottom-contact poly(3,3‴-didodecylquaterthiophene) thin-film transistors with gold source-drain electrodes modified by alkanethiol monolayers. Langmuir. 2008, 24,(20), 11889-11894.
  2. Anne Kathrena A.Aliganga; Anne-Sophie Duwez; Silvia Mittler. Binary mixtures of self-assembled monolayers of 1,8-octanedithiol and 1-octanethiol for a controlled growth of gold nanoparticles. Organic Electronics. 2006, 7,(5), 337-350.