2-Ethylanthraquinone, 98%
2-Ethylanthraquinone, 98%
2-Ethylanthraquinone, 98%
Thermo Scientific Chemicals

2-Ethylanthraquinone, 98%

CAS: 84-51-5 | C16H12O2 | 236.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number L07206.22
also known as L07206-22
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.90
Save 7.25 (15%)
Each
Chemical Identifiers
CAS84-51-5
IUPAC Name2-ethyl-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC16H12O2
InChI KeySJEBAWHUJDUKQK-UHFFFAOYSA-N
SMILESCCC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)106.0-112.0?C
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥97.5%
Appearance (Color)Yellow
2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A.Drelinkiewicz; M.Hasik; M.Kloc. Pd/polyaniline as the catalysts for 2-ethylanthraquinone hydrogenation. The effect of palladium dispersion. Catalysis Letters. 2000, 64,(1), 41-47.
  2. E.Santacesaria; M. Di Serio; R.Velotti; U.Leone. Hydrogenation of the aromatic rings of 2-ethylanthraquinone on palladium catalyst. Journal of Molecular Catalysis. 1994, 94,(1), 37-46.