1,2-Epoxydecane, 97%
1,2-Epoxydecane, 97%
1,2-Epoxydecane, 97%
1,2-Epoxydecane, 97%
Thermo Scientific Chemicals

1,2-Epoxydecane, 97%

CAS: 2404-44-6 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07289.06
also known as L07289-06
Price (USD)
34.65
Online Exclusive
38.20
Save 3.55 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
38.20
Save 3.55 (9%)
Each
Add to cart
Chemical Identifiers
CAS2404-44-6
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4265-1.4315 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only