4-Fluorobenzenesulfonyl chloride, 98%
4-Fluorobenzenesulfonyl chloride, 98%
4-Fluorobenzenesulfonyl chloride, 98%
4-Fluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-Fluorobenzenesulfonyl chloride, 98%

CAS: 349-88-2 | C6H4ClFO2S | 194.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07299.14
also known as L07299-14
Price (USD)
60.65
Online Exclusive
67.10
Save 6.45 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
67.10
Save 6.45 (10%)
Each
Add to cart
Chemical Identifiers
CAS349-88-2
IUPAC Name4-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClFO2S
InChI KeyBFXHJFKKRGVUMU-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream to pale brown or pale yellow
Melting Point (clear melt)33-38°C
FormPowder or crystalline powder or crystals or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only