Methyl 4-chloro-3-nitrobenzoate, 98%
Methyl 4-chloro-3-nitrobenzoate, 98%
Methyl 4-chloro-3-nitrobenzoate, 98%
Methyl 4-chloro-3-nitrobenzoate, 98%
Thermo Scientific Chemicals

Methyl 4-chloro-3-nitrobenzoate, 98%

CAS: 14719-83-6 | C8H6ClNO4 | 215.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07407.06
also known as L07407-06
Price (USD)
54.65
Online Exclusive
60.70
Save 6.05 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
54.65
Online Exclusive
60.70
Save 6.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS14719-83-6
IUPAC Namemethyl 4-chloro-3-nitrobenzoate
Molecular FormulaC8H6ClNO4
InChI KeyXRTKWPWDSUNLHS-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to cream
Melting Point (clear melt)77.5-83.5?C
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. M. K. ChantooniJr.; I. M. Kolthoff. Resolution of acid strength in tert-butyl alcohol and isopropyl alcohol of substituted benzoic acids, phenols, and aliphatic carboxylic acids. Anal. Chem. 1979, 51, (1), 133-140.
  2. Joseph Miller. The SN Mechanism in Aromatic Compounds. VI. Carbonyl and Nitrile Substituents. J. Am. Chem. Soc. 1954, 76, (2), 448-450.