Ethidium bromide, 95%
Ethidium bromide, 95%
Ethidium bromide, 95%
Thermo Scientific Chemicals

Ethidium bromide, 95%

CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07482.03
also known as L07482-03
Price (USD)
39.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.70
Each
Chemical Identifiers
CAS1239-45-8
IUPAC Name3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Molecular FormulaC21H20BrN3
InChI KeyZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to dark red to purple to dark purple
Assay (Argentometric Titration)≥94.0 to ≤106.0% (non-U.S. specification)
Loss on Drying≤5.0% (non-U.S. specification)
Assay from Supplier's CofA≥94.0% (U.S. specification)
FormCrystals or powder or crystalline powder
For DNA isolation and visualization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only