4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%
Thermo Scientific Chemicals

4-Amino-6-hydroxy-2-methylpyrimidine dihydrate, 98+%

CAS: 388582-41-0 | C5H11N3O3 | 161.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07496.06
also known as L07496-06
Price (USD)
37.65
Online Exclusive
41.70
Save 4.05 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.70
Save 4.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS388582-41-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only