2,5-Dichlorobenzyl chloride, 97%
2,5-Dichlorobenzyl chloride, 97%
2,5-Dichlorobenzyl chloride, 97%
Thermo Scientific Chemicals

2,5-Dichlorobenzyl chloride, 97%

CAS: 2745-49-5 | C7H5Cl3 | 195.467 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07506.14
also known as L07506-14
Price (USD)
307.65
Online Exclusive
342.00
Save 34.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
307.65
Online Exclusive
342.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS2745-49-5
IUPAC Name1,4-dichloro-2-(chloromethyl)benzene
Molecular FormulaC7H5Cl3
InChI KeyOMZINLIPPVNUOG-UHFFFAOYSA-N
SMILESClCC1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5725-1.5775 @ 20°C
FormLiquid
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
RUO – Research Use Only