Acetazolamide, 99%
Acetazolamide, 99%
Acetazolamide, 99%
Thermo Scientific Chemicals

Acetazolamide, 99%

CAS: 59-66-5 | C4H6N4O3S2 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07562.06
also known as L07562-06
Price (USD)
40.30
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.30
Each
Chemical Identifiers
CAS59-66-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥98.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Acetazolamide acts as a carbonic anhydrase inhibitor which increases cerebral blood flow. It inhibits water permeability of membranes by interacting with aquaporins. It is used for the medical treatment of glaucoma, epileptic seizure, idiopathic intracranial hypertension, altitude sickness, cystinuria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Junwei Gao; Xiaohua Wang; Yongjie Chang; Jianzhao Zhang; Qianliu Song; Heming Yu; Xuejun Li. Acetazolamide inhibits osmotic water permeability by interaction with aquaporin-1.Analytical Biochemistry.2006, 350 (2), 165-170 .
  2. S. Nigel Cox, FRCS; Elizabeth Hay, MD; Alan C. Bird, MD. Treatment of Chronic Macular Edema With Acetazolamide.Arch Ophthalmol.1988, 106 (9), 1190-1195.