1,2-Epoxyhexane, 96%
1,2-Epoxyhexane, 96%
1,2-Epoxyhexane, 96%
1,2-Epoxyhexane, 96%
Thermo Scientific Chemicals

1,2-Epoxyhexane, 96%

CAS: 1436-34-6 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07609.14
also known as L07609-14
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.30
Save 8.65 (14%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.30
Save 8.65 (14%)
Each
Add to cart
Chemical Identifiers
CAS1436-34-6
IUPAC Name2-butyloxirane
Molecular FormulaC6H12O
InChI KeyWHNBDXQTMPYBAT-UHFFFAOYNA-N
SMILESCCCCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4035-1.4095 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥95.0%
FormLiquid
Identification (FTIR)Conforms
1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Joseph M. Ready.; Eric N. Jacobsen. Highly Active Oligomeric (salen)Co Catalysts for Asymmetric Epoxide Ring-Opening Reactions. J. Am. Chem. Soc. 2001, 123, (11), 2687-2688.
  2. James H. Babler.; William E. Bauta. Cyclization of 6-bromo-1,2-epoxyhexane promoted by metal-halogen exchange: unexpected regioselectivity. Tetrahedron Letters. 1984, 25, (39), 4323-4324.