1,7-Octadiene, 97%
1,7-Octadiene, 97%
1,7-Octadiene, 97%
Thermo Scientific Chemicals

1,7-Octadiene, 97%

CAS: 3710-30-3 | C8H14 | 110.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L07659.AP
also known as L07659-AP
Price (USD)
248.65
Online Exclusive
276.00
Save 27.35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
248.65
Online Exclusive
276.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS3710-30-3
IUPAC Nameocta-1,7-diene
Molecular FormulaC8H14
InChI KeyXWJBRBSPAODJER-UHFFFAOYSA-N
SMILESC=CCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear,colourless to pale yellow
FormLiquid
Refractive Index1.4200-1.4250 @20?C
Assay (GC)>96.0%
1,7-Octadiene is used in organic synthesis. It is also used to study and assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes. Further, it is used in a study to investigate micropatterned surfaces prepared by plasma polymerization. It acts as a crosslinker as well as a source of ethylene in cross-enyne metathesis (CEYM)-related reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Octadiene is used in organic synthesis. It is also used to study and assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes. Further, it is used in a study to investigate micropatterned surfaces prepared by plasma polymerization. It acts as a crosslinker as well as a source of ethylene in cross-enyne metathesis (CEYM)-related reactions.

Solubility
Miscible with methanol and cyclohexane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Bykov, V. I.; Chernova, O. B.; Belyaev, B. A.; Butenko, T. A.; Finkelshtein, E. S. Stereochemical features of 1,7-octadiene metathesis on the MoCl5/SiO2-Me4Sn catalytic system. Pet. Chem. 2015, 55 (7), 549-551.
  2. Apisuk, W.; Nomura, K. Efficient Terpolymerization of Ethylene and Styrene with 1,7-Octadiene by Aryloxo Modified Half-Titanocenes-Cocatalyst Systems: Efficient Introduction of the Reactive Functionality. Macromol. Chem. Phys. 2014, 215 (18), 1785-1791.