D-Lysine monohydrochloride, 98%
D-Lysine monohydrochloride, 98%
D-Lysine monohydrochloride, 98%
D-Lysine monohydrochloride, 98%
Thermo Scientific Chemicals

D-Lysine monohydrochloride, 98%

CAS: 7274-88-6 | C6H15ClN2O2 | 182.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07710.03
also known as L07710-03
Price (USD)
36.65
Online Exclusive
41.00
Save 4.35 (11%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
41.00
Save 4.35 (11%)
Each
Add to cart
Chemical Identifiers
CAS7274-88-6
IUPAC Name(2R)-2,6-diaminohexanoic acid hydrochloride
Molecular FormulaC6H15ClN2O2
InChI KeyBVHLGVCQOALMSV-XZNNNFJINA-N
SMILESCl.NCCCC[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder
Optical Rotation-20.5 ? 1.5? (c=5 in 5N HCl)
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (Dry wt. basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only