Benzoyleneurea, 98%
Benzoyleneurea, 98%
Benzoyleneurea, 98%
Thermo Scientific Chemicals

Benzoyleneurea, 98%

CAS: 86-96-4 | C8H6N2O2 | 162.148 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07763.06
also known as L07763-06
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.90
Save 5.25
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.90
Save 5.25
Each
Chemical Identifiers
CAS86-96-4
IUPAC Name1,2,3,4-tetrahydroquinazoline-2,4-dione
Molecular FormulaC8H6N2O2
InChI KeySDQJTWBNWQABLE-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)White to cream or pink to gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only