1-Naphthoic hydrazide, 98+%
1-Naphthoic hydrazide, 98+%
1-Naphthoic hydrazide, 98+%
Thermo Scientific Chemicals

1-Naphthoic hydrazide, 98+%

CAS: 43038-45-5 | C11H10N2O | 186.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07942.14
also known as L07942-14
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS43038-45-5
IUPAC Namenaphthalene-1-carbohydrazide
Molecular FormulaC11H10N2O
InChI KeyVMFUMDXVTKTZQY-UHFFFAOYSA-N
SMILESNNC(=O)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only