Ethyl 3-(4-methoxybenzoyl)propionate, 98%
Ethyl 3-(4-methoxybenzoyl)propionate, 98%
Ethyl 3-(4-methoxybenzoyl)propionate, 98%
Ethyl 3-(4-methoxybenzoyl)propionate, 98%
Thermo Scientific Chemicals

Ethyl 3-(4-methoxybenzoyl)propionate, 98%

CAS: 15118-67-9 | C13H16O4 | 236.267 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07989.06
also known as L07989-06
Price (USD)
66.65
Online Exclusive
73.80
Save 7.15 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
73.80
Save 7.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS15118-67-9
IUPAC Nameethyl 4-(4-methoxyphenyl)-4-oxobutanoate
Molecular FormulaC13H16O4
InChI KeyFYUAOZFEVHSJTO-UHFFFAOYSA-N
SMILESCCOC(=O)CCC(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to cream
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)48.0-54.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only