2-Methyl-3-pentanol, 97+%
2-Methyl-3-pentanol, 97+%
2-Methyl-3-pentanol, 97+%
Thermo Scientific Chemicals

2-Methyl-3-pentanol, 97+%

CAS: 565-67-3 | C6H14O | 102.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L08015.18
also known as L08015-18
Price (USD)
320.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
320.00
Each
Chemical Identifiers
CAS565-67-3
IUPAC Name2-methylpentan-3-ol
Molecular FormulaC6H14O
InChI KeyISTJMQSHILQAEC-UHFFFAOYNA-N
SMILESCCC(O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Refractive Index1.4145-1.4195 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.

Solubility
Amalgamate with ethanol, ethyl ether, benzene. Insoluble in water

Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only
Angelo C Salvador; Inês Baptista; António S Barros; Newton C M Gomes; Angela Cunha; Adelaide Almeida; Silvia M Rocha. Can volatile organic metabolites be used to simultaneously assess microbial and mite contamination level in cereal grains and coffee beans?PLoS ONE.1968, 8 (4), e59338.