Thermo Scientific Chemicals

3-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L08072.06
5 g, Each
Thermo Scientific Chemicals

3-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L08072.06
5 g, Each
Quantity
Catalog number: L08072.06
also known as L08072-06
Price (USD)
Price: 121.00
Online price: 102.65
Your price:
Quantity
-

Chemical Identifiers

CAS
100-80-1
IUPAC Name
1-ethenyl-3-methylbenzene
Molecular Formula
C9H10
InChI Key
JZHGRUMIRATHIU-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C=C)=C1
Form
Liquid
Assay (GC)
>97.5%
Refractive Index
1.5390-1.5430 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Solution Test
Clear (10% in Diethyl ether)

Description

3-Methylstyreneis used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylstyreneis used as pharmaceutical intermediates.

Solubility
Soluble in Methanol, Ether, Benzene, Acetone, Ethanol, Heptane. Soluble in water (0.09 g/L) at 20°C.

Notes
Store at 4°C. Pr
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text