trans-2-Methyl-2-butenal, 97%
trans-2-Methyl-2-butenal, 97%
trans-2-Methyl-2-butenal, 97%
Thermo Scientific Chemicals

trans-2-Methyl-2-butenal, 97%

CAS: 497-03-0 | C5H8O | 84.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08113.06
also known as L08113-06
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
49.70
Save 7.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
49.70
Save 7.05
Each
Chemical Identifiers
CAS497-03-0
IUPAC Name(2E)-2-methylbut-2-enal
Molecular FormulaC5H8O
InChI KeyACWQBUSCFPJUPN-HWKANZROSA-N
SMILESC\C=C(/C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.4455-1.4505 @ 20?C
Free acid (titration)≤3.0%
Assay (GC)≥96.0%
Identification (FTIR)Conforms
View more
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Elena Jiménez.; Beatriz Lanza.; María Antiñolo.; José Albaladejo. Influence of temperature on the chemical removal of 3-methylbutanal, trans-2-methyl-2-butenal, and 3-methyl-2-butenal by OH radicals in the troposphere. Atmospheric Environment. 2009, 43, (26), 4043-4049.
  2. Daryl D. Rowan.; Heather P. Lane.; John M. Allen.; Simon Fielder.; Martin B. Hunt. Biosynthesis of 2-Methylbutyl, 2-Methyl-2-butenyl, and 2-Methylbutanoate Esters in Red Delicious and Granny Smith Apples Using Deuterium-Labeled Substrates. J. Agric. Food Chem. 1996, 44, (10), 3276-3285.