Calcium DL-glycerate dihydrate
Calcium DL-glycerate dihydrate
Calcium DL-glycerate dihydrate
Thermo Scientific Chemicals

Calcium DL-glycerate dihydrate

CAS: 67525-74-0 | C3H5O4 | 105.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08214.03
also known as L08214-03
Price (USD)
78.65
Online Exclusive
87.00
Save 8.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
78.65
Online Exclusive
87.00
Save 8.35 (10%)
Each
Chemical Identifiers
CAS67525-74-0
IUPAC Name(2R)-2,3-dihydroxypropanoate
Molecular FormulaC3H5O4
InChI KeyRBNPOMFGQQGHHO-UWTATZPHSA-M
SMILESOC[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in cold and hot water.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Tooru Taga; Makoto Ohashi; Kenji Osaki. Interactions of Calcium Ions with Carbohydrates: Crystal Structure of Calcium DL-Glycerate Dihydrate. Bulletin of the Chemical Society of Japan. 1978, 51 (6), 1697-1700.