Specifications
Beilstein519794
CAS7073-36-1
Chemical Name or Material2-Chloro-4-nitrobenzoyl chloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECS Number230-367-4
View more