4-tert-Butylcyclohexanecarboxylic acid, predominantly trans, 98+%
4-tert-Butylcyclohexanecarboxylic acid, predominantly trans, 98+%
4-tert-Butylcyclohexanecarboxylic acid, predominantly trans, 98+%
Thermo Scientific Chemicals

4-tert-Butylcyclohexanecarboxylic acid, predominantly trans, 98+%

CAS: 5451-55-8 | C11H20O2 | 184.279 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08248.06
also known as L08248-06
Price (USD)
56.65
Online Exclusive
62.70
Save 6.05 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.70
Save 6.05 (10%)
Each
Chemical Identifiers
CAS5451-55-8
IUPAC Name4-tert-butylcyclohexane-1-carboxylic acid
Molecular FormulaC11H20O2
InChI KeyQVQKEGYITJBHRQ-UHFFFAOYSA-N
SMILESCC(C)(C)C1CCC(CC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only