Hydroquinonesulfonic acid potassium salt, 98+%
Hydroquinonesulfonic acid potassium salt, 98+%
Hydroquinonesulfonic acid potassium salt, 98+%
Thermo Scientific Chemicals

Hydroquinonesulfonic acid potassium salt, 98+%

CAS: 21799-87-1 | C6H5KO5S | 228.259 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L08406.22
also known as L08406-22
Price (USD)
40.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
40.00
Each
Chemical Identifiers
CAS21799-87-1
IUPAC Namecalcium bis(2,5-dihydroxybenzene-1-sulfonate)
Molecular FormulaC12H10CaO10S2
InChI KeyQGNBTYAQAPLTMX-UHFFFAOYSA-L
SMILES[Ca++].OC1=CC=C(O)C(=C1)S([O-])(=O)=O.OC1=CC=C(O)C(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥98.0%
FormCrystals or powder or crystalline powder
It is employed in the synthesis and properties of sulfonated polyarylene ether nitrile copolymers for PEM with high thermal stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and properties of sulfonated polyarylene ether nitrile copolymers for PEM with high thermal stability.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Jian Lin, et al. Novel wholly aromatic lyotropic liquid crystalline polyesters. Synthesis, characterization, and solution properties.Macromolecules.,1992,25(26), 7107-7113.
  2. Lan Chen, et al. Synthesis and properties of sulfonated polyarylene ether nitrile copolymers for PEM with high thermal stability.J. Polym. Res.,2013,20(45).